Ethyl-4-hydroxy-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate 97%
Supplier: Apollo Scientific
Warning
PC5849-1G PC5849-250MG PC5849-5G
APOSPC5849-1GEA 103 EUR
APOSPC5849-1G APOSPC5849-250MG APOSPC5849-5G
Ethyl-4-hydroxy-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate 97%
Ethyl-4-hydroxy-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
Formula: C₁₃H₁₀F₃NO₃S
MW: 317.29 g/mol
Melting Pt: 114…116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00816796
CAS Number: 240800-53-7