2-Fluoro-5-(trifluoromethoxy)benzoic acid 98%
Supplier: Apollo Scientific
Warning
PC5896-5G PC5896-1G PC5896-10G
APOSPC5896-5GEA 85.7 EUR
APOSPC5896-5G APOSPC5896-1G APOSPC5896-10G
2-Fluoro-5-(trifluoromethoxy)benzoic acid 98%
2-Fluoro-5-(trifluoromethoxy)benzoic acid
Formula: C₈H₄F₄O₃
MW: 224.11 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD04115887
CAS Number: 886497-85-4