2-(Perfluorobutyl)ethyl acrylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Acrylate, Acrylic acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl ester, Acrylic acid 1H,1H,2H,2H-Nonafluorohexyl ester, 1H,1H,2H,2H-Nonafluorohexyl Acrylate
PC5906D-5G PC5906D-500MG
APOSPC5906D-5GEA 55.1 EUR
APOSPC5906D-5G APOSPC5906D-500MG
2-(Perfluorobutyl)ethyl acrylate 98%
2-(Perfluorobutyl)ethyl acrylate
Formula: C₉H₇F₉O₂
Boiling Pt: 160 °C
Melting Pt: –68 °C
Density: 1.44 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00236104
CAS Number: 52591-27-2
UN: 3272
ADR: 3,III