2-Fluoro-p-cresol
Supplier: Apollo Scientific
Danger
Synonyms: 2-Fluoro-4-methylphenol, 1-Fluoro-2-hydroxy-5-methylbenzene
PC5944-5G PC5944-100G PC5944-25G
APOSPC5944-5GEA 24.8 EUR
APOSPC5944-5G APOSPC5944-100G APOSPC5944-25G
2-Fluoro-p-cresol
2-Fluoro-p-cresol
Formula: C₇H₇FO
Boiling Pt: 172…174 °C
Density: 1.155 g/cm³ (25 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD03094332
CAS Number: 452-81-3