1-Fluoro-2-ethynylbenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: (2-Fluorophenyl)ethyne, 1-Ethynyl-2-fluorobenzene, (2-Fluorophenyl)acetylene
PC5957-5G PC5957-1G PC5957-25G
APOSPC5957-5GEA 34.8 EUR
APOSPC5957-5G APOSPC5957-1G APOSPC5957-25G
1-Fluoro-2-ethynylbenzene 97%
1-Fluoro-2-ethynylbenzene
Formula: C₈H₅F
Boiling Pt: 150 °C
Density: 1.06 g/cm³ (25 °C)
Flash Pt: 37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00799540
CAS Number: 766-49-4
UN: 1993
ADR: 3,III