1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol 96%
Supplier: Apollo Scientific
Warning
Synonyms: 1H,1H,12H,12H-Perfluoro-1,12-dodecanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Eicosafluoro-1,12-dodecandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-Icosafluorododecane-1,12-diol, 1H,1H,12H,12H-Icosafluoro-1,12-dodecanediol, 1H,1H,12H,12H-Perfluoro-1,12-dodecandiol
PC5993-25G PC5993-5G PC5993-1G
APOSPC5993-25GEA 319 EUR
APOSPC5993-25G APOSPC5993-5G APOSPC5993-1G
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol 96%
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol
Formula: C₁₂H₆F₂₀O₂
MW: 562.15 g/mol
Boiling Pt: 170 °C/5mmHg
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00236625
CAS Number: 183162-43-8