3-Bromo-6-chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Warning
PC6019-1G PC6019-500MG
APOSPC6019-1GEA 386 EUR
APOSPC6019-1G APOSPC6019-500MG
3-Bromo-6-chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine
3-Bromo-6-chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine
Formula: C₈H₃BrClF₃N₂
MW: 299.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025923
CAS Number: 1160474-82-7