2-Fluoro-5-(hydroxymethyl)benzonitrile
Supplier: Apollo Scientific
Warning
PC6132-1G PC6132-5G PC6132-25G
APOSPC6132-1GEA 21.9 EUR
APOSPC6132-1G APOSPC6132-5G APOSPC6132-25G
2-Fluoro-5-(hydroxymethyl)benzonitrile
2-Fluoro-5-(hydroxymethyl)benzonitrile
Formula: C₈H₆FNO
MW: 151.14 g/mol
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD03094340
CAS Number: 227609-85-0