6-Chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine
Supplier: Apollo Scientific
PC6151-1G PC6151-500MG
APOSPC6151-1GEA 207 EUR
APOSPC6151-1G APOSPC6151-500MG
6-Chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine
6-Chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine
Formula: C₈H₄ClF₃N₂
MW: 220.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04619561
CAS Number: 189116-20-9