Ethyl 3-bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Supplier: Apollo Scientific
Warning
PC6154-1G PC6154-500MG
APOSPC6154-1GEA 134 EUR
APOSPC6154-1G APOSPC6154-500MG
Ethyl 3-bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₁H₈BrF₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12025928
CAS Number: 1160474-86-1