1-Bromoperfluorooctane 98%
Supplier: Apollo Scientific
Warning
Synonyms: Perflubron, 1-Bromoheptadecafluorooctane, Perfluorooctyl bromide
PC6167-100G PC6167-25G PC6167-5G
APOSPC6167-100GEA 197 EUR
APOSPC6167-100G APOSPC6167-25G APOSPC6167-5G
1-Bromoperfluorooctane 98%
1-Bromoperfluorooctane
Formula: BrF₂C(CF₂)₆CF₃
Boiling Pt: 141…143 °C
Melting Pt: 6 °C
Density: 1.93 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042082
CAS Number: 423-55-2
EINECS: 207-028-4