α,α,α,2,6-Pentafluorotoluene
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Difluorobenzotrifluoride
PC6200-25G PC6200-5G PC6200-100G
APOSPC6200-25GEA 207 EUR
APOSPC6200-25G APOSPC6200-5G APOSPC6200-100G
α,α,α,2,6-Pentafluorotoluene
α,α,α,2,6-Pentafluorotoluene
Formula: C₇H₃F₅
Boiling Pt: 106…107 °C
Storage Temperature: Ambient
MDL Number: MFCD03412213
CAS Number: 64248-60-8
UN: 1993
ADR: 3,III