Perfluorophenanthrene 80%

Supplier: Apollo Scientific
Warning

Synonyms: Perfluoroperhydrophenanthrene

PC6202-250G PC6202-50G
APOSPC6202-250GEA 286 EUR
APOSPC6202-250G APOSPC6202-50G
Perfluorophenanthrene 80%
Perfluorophenanthrene
Formula: C₁₄F₂₄
MW: 624.116 g/mol
Boiling Pt: 215 °C
Melting Pt: –20 °C
Density: 2.03 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042596
CAS Number: 306-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR