Diethyl-4,4-difluoroheptanedioate 96%
Supplier: Apollo Scientific
Warning
PC6217-1G PC6217-5G PC6217-25G
APOSPC6217-1GEA 0 EUR
APOSPC6217-1G APOSPC6217-5G APOSPC6217-25G
Diethyl-4,4-difluoroheptanedioate 96%
Diethyl-4,4-difluoroheptanedioate
Formula: C₁₁H₁₈F₂O₄
Boiling Pt: 108 °C/3mmHg
Density: 1.1 g/cm³ (25 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD08146634
CAS Number: 22515-16-8