Potassium hexafluorosilicate 98%
Supplier: Apollo Scientific
Danger
Synonyms: Potassium fluorosilicate
PC6294-1KG PC6294-5KG
APOSPC6294-1KGEA 227 EUR
APOSPC6294-1KG APOSPC6294-5KG
Potassium hexafluorosilicate 98%
Potassium hexafluorosilicate
Formula: F₆K₂Si
Density: 2.665 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040666
CAS Number: 16871-90-2
UN: 2655
ADR: 6.1,III