4-(Difluoromethoxy)phenylacetonitrile 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(4-(Difluoromethoxy)phenyl)acetonitrile
PC6323-10G PC6323-1G PC6323-5G PC6323-25G
APOSPC6323-10GEA 575 EUR
APOSPC6323-10G APOSPC6323-1G APOSPC6323-5G APOSPC6323-25G
4-(Difluoromethoxy)phenylacetonitrile 98%
4-(Difluoromethoxy)phenylacetonitrile
Formula: C₉H₇F₂NO
MW: 183.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236242
CAS Number: 41429-16-7
UN: 3276
ADR: 6.1 III