4-Chloro-3-fluoropyridine
Supplier: Apollo Scientific
Warning
PC6447-25G PC6447-5G PC6447-1G
APOSPC6447-25GEA 126 EUR
APOSPC6447-25G APOSPC6447-5G APOSPC6447-1G
4-Chloro-3-fluoropyridine
4-Chloro-3-fluoropyridine
Formula: C₅H₃ClFN
MW: 131.54 g/mol
Boiling Pt: 138…143 °C
Melting Pt: –27…–24 °C
Density: 1.333 g/cm³ (25 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD03453233
CAS Number: 2546-56-7
UN: 1993
ADR: 3 III