Sodium fluoride 99.99%
Supplier: Apollo Scientific
Danger
PC6466B-1G PC6466B-25G
APOSPC6466B-1GEA 22 EUR
APOSPC6466B-1G APOSPC6466B-25G
Sodium fluoride 99.99%
Sodium fluoride
Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1700
Melting Pt: 988 °C
Density: 2.78 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1 III
Merck Index: 13,08691