2,3-Difluorotoluene 97%
Supplier: Apollo Scientific
Danger
PC6523-100G PC6523-25G PC6523-5G
APOSPC6523-100GEA 82.8 EUR
APOSPC6523-100G APOSPC6523-25G APOSPC6523-5G
2,3-Difluorotoluene 97%
2,3-Difluorotoluene
Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 119…121 °C
Density: 1.096 g/cm³ (25 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD01631506
CAS Number: 3828-49-7
UN: 1993
ADR: 3 II