3-Amino-4-(methylthio)benzotrifluoride 97%
Supplier: Apollo Scientific
Synonyms: 2-Methylthio-5-(trifluoromethyl)aniline
PC6554-25G PC6554-5G
APOSPC6554-25GEA 553 EUR
APOSPC6554-25G APOSPC6554-5G
3-Amino-4-(methylthio)benzotrifluoride 97%
3-Amino-4-(methylthio)benzotrifluoride
Formula: C₈H₈F₃NS
Boiling Pt: 111…112 °C/12mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052715
CAS Number: 207974-07-0