ω,ω-Difluoroacetophenone 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2-Difluoroacetophenone, 2,2-Difluoro-1-phenylethanone, α,α-Difluoroacetophenone
PC6686-1G PC6686-5G
APOSPC6686-1GEA 71.2 EUR
APOSPC6686-1G APOSPC6686-5G
ω,ω-Difluoroacetophenone 95%
ω,ω-Difluoroacetophenone
Formula: C₈H₆F₂O
MW: 156.13 g/mol
Boiling Pt: 63…65 °C/11mmHg
Melting Pt: 9 °C
Storage Temperature: Ambient
MDL Number: MFCD06246879
CAS Number: 395-01-7
UN: 1993
ADR: 3 III