4-(1,1,2,2-Tetrafluoroethoxy)aniline 98%
Supplier: Apollo Scientific
|
Danger
|
PC6762A-1G
PC6762A-5G
APOSPC6762A-1GEA
111
EUR
APOSPC6762A-1G
APOSPC6762A-5G
4-(1,1,2,2-Tetrafluoroethoxy)aniline 98%
4-(1,1,2,2-Tetrafluoroethoxy)aniline
Formula:
C₈H₇F₄NO Boiling Pt: 104…108 °C/11mmHg Density: 1.379 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042377 CAS Number: 713-62-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...