4-(Trifluoromethoxy)benzenesulphonamide
Supplier: Apollo Scientific
Warning
PC6842-1G PC6842-5G
APOSPC6842-1GEA 14.7 EUR
APOSPC6842-1G APOSPC6842-5G
4-(Trifluoromethoxy)benzenesulphonamide
4-(Trifluoromethoxy)benzenesulphonamide
Formula: C₇H₆F₃NO₃S
MW: 241.19 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00798381
CAS Number: 1513-45-7