2-Chloro-5-fluoro-4-methoxypyrimidine 98%
Supplier: Apollo Scientific
Warning
PC6888-5G PC6888-25G
APOSPC6888-5GEA 69.6 EUR
APOSPC6888-5G APOSPC6888-25G
2-Chloro-5-fluoro-4-methoxypyrimidine 98%
2-Chloro-5-fluoro-4-methoxypyrimidine
Formula: C₅H₄ClFN₂O
Melting Pt: 37…39 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07440062
CAS Number: 37554-70-4
UN: 2811
ADR: 6.1,III