1,1,2-Trichloro-3,3,3-trifluoropropene 97%
Supplier: Apollo Scientific
Synonyms: 1,1,2-Trichlorotrifluoropropene, 1,1,2-Trichloro-3,3,3-trifluoro-1-propene
PC7090-25G PC7090-5G
APOSPC7090-25GEA 323 EUR
APOSPC7090-25G APOSPC7090-5G
1,1,2-Trichloro-3,3,3-trifluoropropene 97%
1,1,2-Trichloro-3,3,3-trifluoropropene
Formula: C₃Cl₃F₃
MW: 199.39 g/mol
Boiling Pt: 88 °C
Density: 1.617 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013672
CAS Number: 431-52-7