2',4',5'-Trifluoroacetophenone 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2,4,5-Trifluorophenyl)ethan-1-one, 2,4,5-Trifluoroacetophenone
PC7202J-100G PC7202J-25G PC7202J-5G
APOSPC7202J-100GEA 296 EUR
APOSPC7202J-100G APOSPC7202J-25G APOSPC7202J-5G
2',4',5'-Trifluoroacetophenone 99%
2',4',5'-Trifluoroacetophenone
Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 99…101 °C/20mmHg
Melting Pt: –7 °C
Density: 1.331 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00061193
CAS Number: 129322-83-4