1-(4-Fluorophenyl)-2-(methylsulphonyl)-1-ethanone
Supplier: Apollo Scientific
Warning
Synonyms: 4'-Fluoro-2-(methylsulphonyl)acetophenone
PC7223-10G PC7223-1G PC7223-5G
APOSPC7223-10GEA 531 EUR
APOSPC7223-10G APOSPC7223-1G APOSPC7223-5G
1-(4-Fluorophenyl)-2-(methylsulphonyl)-1-ethanone
1-(4-Fluorophenyl)-2-(methylsulphonyl)-1-ethanone
Formula: C₉H₉FO₃S
MW: 216.23 g/mol
Melting Pt: 111…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00202594
CAS Number: 82652-17-3