4'-Methyl-2,2,2-trifluoroacetophenone 96%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2,2-Trifluoro-1-(p-tolyl)ethanone, 4-(Trifluoroacetyl)toluene, 2,2,2-Trifluoro-1-(4-methylphenyl)ethan-1-one
PC7250-1G PC7250-25G PC7250-5G
APOSPC7250-1GEA 21.9 EUR
APOSPC7250-1G APOSPC7250-25G APOSPC7250-5G
4'-Methyl-2,2,2-trifluoroacetophenone 96%
4'-Methyl-2,2,2-trifluoroacetophenone
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 179…180 °C
Melting Pt: 44289 °C
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00052338
CAS Number: 394-59-2