2,4,5-Trifluoroaniline 98%
Supplier: Apollo Scientific
Warning
PC7259-100G PC7259-25G PC7259-5G
APOSPC7259-100GEA 62.5 EUR
APOSPC7259-100G APOSPC7259-25G APOSPC7259-5G
2,4,5-Trifluoroaniline 98%
2,4,5-Trifluoroaniline
Formula: C₆H₄F₃N
Melting Pt: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00007649
CAS Number: 367-34-0
EINECS: 206-692-2