2,3,6-Trifluorobenzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC7265G-25G PC7265G-5G
APOSPC7265G-25GEA 132 EUR
APOSPC7265G-25G APOSPC7265G-5G
2,3,6-Trifluorobenzaldehyde 97%
2,3,6-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 159 °C
Density: 1.425 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00061195
CAS Number: 104451-70-9