4-Bromo-2,3-difluoroanisole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-2,3-difluoro-4-methoxybenzene
PC7278-5G PC7278-100G PC7278-25G
APOSPC7278-5GEA 37.7 EUR
APOSPC7278-5G APOSPC7278-100G APOSPC7278-25G
4-Bromo-2,3-difluoroanisole 98%
4-Bromo-2,3-difluoroanisole
Formula: C₇H₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD07368017
CAS Number: 406482-22-2