2,3,6-Trifluorobenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC7283F-25G PC7283F-5G PC7283F-1G
APOSPC7283F-25GEA 329 EUR
APOSPC7283F-25G APOSPC7283F-5G APOSPC7283F-1G
2,3,6-Trifluorobenzonitrile 98%
2,3,6-Trifluorobenzonitrile
Formula: C₇H₂F₃N
MW: 157.09 g/mol
Boiling Pt: 179 °C
Density: 1.359 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00012287
CAS Number: 136514-17-5
UN: 3276
ADR: 6.1 III