5-Fluoro-2-methoxy-4-pyrimidinone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Fluoro-2-methoxypyrimidin-4(3H)-one
PC7302-1G PC7302-5G PC7302-25G
APOSPC7302-1GEA 21.9 EUR
APOSPC7302-1G APOSPC7302-5G APOSPC7302-25G
5-Fluoro-2-methoxy-4-pyrimidinone 98%
5-Fluoro-2-methoxy-4-pyrimidinone
Formula: C₅H₅FN₂O₂
MW: 144.11 g/mol
Melting Pt: 204…208 °C
Storage Temperature: Ambient
MDL Number: MFCD03265437
CAS Number: 1480-96-2