2,2,2-Trifluoroethyl acrylate 99%
Supplier: Apollo Scientific
Danger
Synonyms: 2,2,2-Trifluoroethylacrylate, Acrylic acid 2,2,2-trifluoroethyl ester
PC7316-100G PC7316-25G
APOSPC7316-100GEA 65.4 EUR
APOSPC7316-100G APOSPC7316-25G
2,2,2-Trifluoroethyl acrylate 99%
2,2,2-Trifluoroethyl acrylate
Formula: C₅H₅F₃O₂
Boiling Pt: 46 °C/125mmHg
Density: 1.216 g/cm³ (25 °C)
Flash Pt: 12 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000444
CAS Number: 407-47-6
UN: 1993
ADR: 3,II