4-Fluoro-3-(trifluoromethoxy)benzaldehyde 95%
Supplier: Apollo Scientific
Warning
PC7363-10G PC7363-1G PC7363-5G
APOSPC7363-10GEA 441 EUR
APOSPC7363-10G APOSPC7363-1G APOSPC7363-5G
4-Fluoro-3-(trifluoromethoxy)benzaldehyde 95%
4-Fluoro-3-(trifluoromethoxy)benzaldehyde
Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660159
CAS Number: 86256-48-6