2-Bromo-5-fluorobenzaldehyde
Supplier: Apollo Scientific
Warning
PC7398-25G PC7398-5G
APOSPC7398-25GEA 39.3 EUR
APOSPC7398-25G APOSPC7398-5G
2-Bromo-5-fluorobenzaldehyde
2-Bromo-5-fluorobenzaldehyde
Formula: C₇H₄BrFO
MW: 203.01 g/mol
Melting Pt: 51…56 °C
Flash Pt: 87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00142872
CAS Number: 94569-84-3