4'-Bromo-2'-(trifluoromethyl)acetanilide
Supplier: Apollo Scientific
Synonyms: N-(4-Bromo-2-(trifluoromethyl)phenyl)acetamide
PC7415-1G PC7415-5G PC7415-25G
APOSPC7415-1GEA 65.4 EUR
APOSPC7415-1G APOSPC7415-5G APOSPC7415-25G
4'-Bromo-2'-(trifluoromethyl)acetanilide
4'-Bromo-2'-(trifluoromethyl)acetanilide
Formula: C₉H₇BrF₃NO
MW: 282.0572 g/mol
MDL Number: MFCD00672936
CAS Number: 29124-62-7