3-(Trifluoromethoxy)aniline 98%
Supplier: Apollo Scientific
Warning
PC7433Z-100G PC7433Z-25G PC7433Z-500G
APOSPC7433Z-100GEA 65.4 EUR
APOSPC7433Z-100G APOSPC7433Z-25G APOSPC7433Z-500G
3-(Trifluoromethoxy)aniline 98%
3-(Trifluoromethoxy)aniline
Formula: C₇H₆F₃NO
Boiling Pt: 72…75 °C/8mmHg
Density: 1.325 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00041511
CAS Number: 1535-73-5
EINECS: 216-256-3