4-(Trifluoromethoxy)anisole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Methoxy-4-(trifluoromethoxy)benzene
PC7434C-100G PC7434C-25G PC7434C-5G
APOSPC7434C-100GEA 106 EUR
APOSPC7434C-100G APOSPC7434C-25G APOSPC7434C-5G
4-(Trifluoromethoxy)anisole 98%
4-(Trifluoromethoxy)anisole
Formula: C₈H₇F₃O₂
MW: 192.14 g/mol
Boiling Pt: 163…165 °C
Density: 1.272 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00216942
CAS Number: 710-18-9
EINECS: 615-239-1
UN: 1993
ADR: 3 III