4-(Trifluoromethoxy)benzaldehyde 95%
Supplier: Apollo Scientific
Warning
PC7434J-100G PC7434J-25G PC7434J-5G
APOSPC7434J-100GEA 84.1 EUR
APOSPC7434J-100G APOSPC7434J-25G APOSPC7434J-5G
4-(Trifluoromethoxy)benzaldehyde 95%
4-(Trifluoromethoxy)benzaldehyde
Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 93 °C/27mmHg
Density: 1.331 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00041530
CAS Number: 659-28-9
EINECS: 211-531-4