4-(Trifluoromethoxy)benzonitrile 98%
Supplier: Apollo Scientific
Warning
PC7438G-100G PC7438G-25G PC7438G-500G
APOSPC7438G-100GEA 78.3 EUR
APOSPC7438G-100G APOSPC7438G-25G APOSPC7438G-500G
4-(Trifluoromethoxy)benzonitrile 98%
4-(Trifluoromethoxy)benzonitrile
Formula: C₈H₄F₃NO
Boiling Pt: 192…193 °C
Density: 1.285 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00039474
CAS Number: 332-25-2
EINECS: 206-363-3
UN: 3276
ADR: 6.1,III