α-Bromo-4-(trifluoromethoxy)toluene 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Bromomethyl)-4-(trifluoromethoxy)benzene, p-(Trifluoromethoxy)benzyl bromide, 4-(Trifluoromethoxy)benzyl bromide
PC7438UB-100G PC7438UB-25G PC7438UB-5G
APOSPC7438UB-100GEA 0 EUR
APOSPC7438UB-100G APOSPC7438UB-25G APOSPC7438UB-5G
α-Bromo-4-(trifluoromethoxy)toluene 97%
α-Bromo-4-(trifluoromethoxy)toluene
Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
Boiling Pt: 82…84 °C/10mmHg
Density: 1.594 g/cm³ (25 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00061238
CAS Number: 50824-05-0
UN: 3261
ADR: 8 II