1-Bromo-2-(trifluoromethoxy)benzene 99%
Supplier: Apollo Scientific
Warning
PC7438UY-100G PC7438UY-500G PC7438UY-50G
APOSPC7438UY-100GEA 48 EUR
APOSPC7438UY-100G APOSPC7438UY-500G APOSPC7438UY-50G
1-Bromo-2-(trifluoromethoxy)benzene 99%
1-Bromo-2-(trifluoromethoxy)benzene
Formula: C₇H₄BrF₃O
Boiling Pt: 158…160 °C
Density: 1.632 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00040942
CAS Number: 64115-88-4
UN: 1993
ADR: 3,III