1-Bromo-3-(trifluoromethoxy)benzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Trifluoromethoxy)bromobenzene
PC7438V-100G PC7438V-500G PC7438V-1KG
APOSPC7438V-100GEA 34.8 EUR
APOSPC7438V-100G APOSPC7438V-500G APOSPC7438V-1KG
1-Bromo-3-(trifluoromethoxy)benzene 99%
1-Bromo-3-(trifluoromethoxy)benzene
Formula: C₇H₄BrF₃O
Boiling Pt: 156…158 °C
Density: 1.62 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00040943
CAS Number: 2252-44-0