1-Iodo-2-(trifluoromethoxy)benzene 98%
Supplier: Apollo Scientific
Warning
PC7439E-100G PC7439E-25G
APOSPC7439E-100GEA 49.3 EUR
APOSPC7439E-100G APOSPC7439E-25G
1-Iodo-2-(trifluoromethoxy)benzene 98%
1-Iodo-2-(trifluoromethoxy)benzene
Formula: C₇H₄F₃IO
MW: 288.01 g/mol
Boiling Pt: 164…165 °C
Density: 1.916 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00042410
CAS Number: 175278-00-9