1-[2-Nitro-4-(trifluoromethyl)phenyl]piperidin-4-ol
Supplier: Apollo Scientific
PC7449-1G PC7449-5G
APOSPC7449-1GEA 106 EUR
APOSPC7449-1G APOSPC7449-5G
1-[2-Nitro-4-(trifluoromethyl)phenyl]piperidin-4-ol
1-[2-Nitro-4-(trifluoromethyl)phenyl]piperidin-4-ol
Formula: C₁₂H₁₃F₃N₂O₃
MW: 290.24 g/mol
Melting Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD04219156
CAS Number: 702650-29-1