2'-(Trifluoromethyl)acetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: o-(Trifluoromethyl)acetophenone, 1-[2-(Trifluoromethyl)phenyl]ethan-1-one, 1-(2-(Trifluoromethyl)phenyl)ethanone
PC7470-25G PC7470-5G
APOSPC7470-25GEA 33.5 EUR
APOSPC7470-25G APOSPC7470-5G
2'-(Trifluoromethyl)acetophenone 98%
2'-(Trifluoromethyl)acetophenone
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 163…164 °C
Melting Pt: 15…17 °C
Density: 1.255 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00000378
CAS Number: 17408-14-9
EINECS: 241-434-2