4-Bromo-3-fluoroaniline 98%
Supplier: Apollo Scientific
Warning
PC7544-100G PC7544-25G PC7544-5G
APOSPC7544-100GEA 114 EUR
APOSPC7544-100G APOSPC7544-25G APOSPC7544-5G
4-Bromo-3-fluoroaniline 98%
4-Bromo-3-fluoroaniline
Formula: C₆H₅BrFN
Melting Pt: 65…70 °C
Density: 0.982 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00672933
CAS Number: 656-65-5