α,α,α-Trifluoro-p-toluonitrile 98%
Supplier: Apollo Scientific
Warning
PC7562-1KG PC7562-250G PC7562-5KG
APOSPC7562-1KGEA 145 EUR
APOSPC7562-1KG APOSPC7562-250G APOSPC7562-5KG
α,α,α-Trifluoro-p-toluonitrile 98%
α,α,α-Trifluoro-p-toluonitrile
Formula: C₈H₄F₃N
Boiling Pt: 183…185 °C
Melting Pt: 37…41 °C
Density: 1.278 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00001826
CAS Number: 455-18-5
EINECS: 207-239-1
UN: 3439
ADR: 6.1,III