α,α,α,2-Tetrafluorotoluene 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Fluoro-2-(trifluoromethyl)benzene, 2-Fluorobenzotrifluoride
PC7596-100G PC7596-500G PC7596-1KG
APOSPC7596-100GEA 21.9 EUR
APOSPC7596-100G APOSPC7596-500G APOSPC7596-1KG
α,α,α,2-Tetrafluorotoluene 97%
α,α,α,2-Tetrafluorotoluene
Formula: C₇H₄F₄
MW: 164.1 g/mol
Boiling Pt: 114…115 °C/750mmHg
Melting Pt: –49 °C
Density: 1.293 g/cm³ (25 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00000374
CAS Number: 392-85-8
EINECS: 206-880-4
UN: 1993
ADR: 3 II